missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Dimethylamino-2-methylpropyl chloride hydrochloride, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 166791000

 View more versions of this product

Catalog No. AC166791000



Specifications

Specifications

3-Dimethylamino-2-methylpropyl chloride hydrochloride
Glass bottle
169°C to 173°C
4261-67-0
100.0
C6H14ClN·HCl
MFCD00012514
1-dimethylamino-2-methyl-3-chloropropane hydrochloride
CC(CN(C)C)CCl.Cl
172.1
172.1
98%
Authentic
White
100g
97.5
98%
(CH3)2NCH2CH(CH3)CH2Cl·HCl
04, IV, 655
SOMIBONUMGNAEP-UHFFFAOYSA-N
3-chloro-N,N,2-trimethylpropan-1-amine;hydrochloride
3083841
Powder

Chemical Identifiers

4261-67-0
172.1
SOMIBONUMGNAEP-UHFFFAOYSA-N
3083841
CC(CN(C)C)CCl.Cl
C6H14ClN·HCl
MFCD00012514
1-dimethylamino-2-methyl-3-chloropropane hydrochloride
3-chloro-N,N,2-trimethylpropan-1-amine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 224-237-6

SDS Documents