Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Dimethylamino-2-methylpropyl chloride hydrochloride, 98%

$31.09 - $327.86

Chemical Identifiers

Chemical Name or Material 3-Dimethylamino-2-methylpropyl chloride hydrochloride
CAS 4261-67-0
Molecular Formula C6H14ClN·HCl
Formula Weight 172.1
MDL Number MFCD00012514
Beilstein 4444785
Synonym 3-Chloro-2-methyl-N,N-dimethylpropylamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0609114

 
alfa aesar™
L0609114
25g Each for $31.09

AAL0609122

 
alfa aesar™
L0609122
100g Each for $88.05

AAL0609136

 
alfa aesar™
L0609136
500g Each for $327.86
Description & Specifications

Specifications

Formula Weight 172.1
Chemical Name or Material 3-Dimethylamino-2-methylpropyl chloride hydrochloride
Molecular Formula C6H14ClN·HCl
MDL Number MFCD00012514
CAS 4261-67-0
Synonym 3-Chloro-2-methyl-N,N-dimethylpropylamine hydrochloride
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Melting Point 167° to 170°C
Beilstein 4444785
Quantity 25g
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.