missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Dimethylamino-1-propanol, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 115881000

 View more versions of this product

Catalog No. AC115881000



Description

Description

Specifications

Specifications

3-Dimethylamino-1-propanol
54°C
Glass bottle
0.886
159.0°C
-35.0°C
99%
98.5
98.5% min. (GC)
(CH3)2N(CH2)3OH
04, I, 433
Solubility in water: soluble
CN(C)CCCO
103.16
103.16
99%
0.8860g/mL
Authentic
1.4350 to 1.4370
6.85 mPa.s (20°C)
Colorless to Yellow
100g
3179-63-3
100.0
C5H13NO
MFCD00002945
1-propanol, 3-dimethylamino, 3-dimethlyamino propanol, 3-dimethylamino propan-1-ol, 3-dimethylamino propanol, 3-dimethylamino-1-propanol, 3-n,n-dimethylamino propanol, dimethylaminopropanol, dimethylpropanolamine, n,n-dimethylpropanolamine
PYSGFFTXMUWEOT-UHFFFAOYSA-N
3-(dimethylamino)propan-1-ol
76646
Liquid

Chemical Identifiers

3179-63-3
103.16
PYSGFFTXMUWEOT-UHFFFAOYSA-N
76646
CN(C)CCCO
C5H13NO
MFCD00002945
1-propanol, 3-dimethylamino, 3-dimethlyamino propanol, 3-dimethylamino propan-1-ol, 3-dimethylamino propanol, 3-dimethylamino-1-propanol, 3-n,n-dimethylamino propanol, dimethylaminopropanol, dimethylpropanolamine, n,n-dimethylpropanolamine
3-(dimethylamino)propan-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 221-659-2

RTECSNumber : UB3155000

TSCA : TSCA

SDS Documents