missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Dimethylamino-1-propanol, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 115880050
Specifications
3-Dimethylamino-1-propanol | |
3179-63-3 | |
100.0 | |
159.0°C | |
Authentic | |
Glass bottle | |
1.4350 to 1.4370 | |
MFCD00002945 | |
04, I, 433 | |
3-dimethylamino-1-propanol, 3-dimethylamino propan-1-ol, 1-propanol, 3-dimethylamino, dimethylaminopropanol, n,n-dimethylpropanolamine, 3-dimethylamino propanol, dimethylpropanolamine, 3-dimethlyamino propanol, 3-n,n-dimethylamino propanol | |
PYSGFFTXMUWEOT-UHFFFAOYSA-N | |
3-(dimethylamino)propan-1-ol | |
76646 | |
103.16 |
99% | |
98.5 | |
0.8860g/mL | |
54°C | |
98.5% min. (GC) | |
C5H13NO | |
(CH3)2N(CH2)3OH | |
5g | |
0.886 | |
Solubility in water: soluble | |
CN(C)CCCO | |
103.17 | |
6.85 mPa.s (20°C) | |
99% |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 221-659-2
RTECSNumber : UB3155000
TSCA : TSCA
RUO â Research Use Only