Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Dimethylamino-1-propanol, 99%

Chemical Identifiers

Chemical Name or Material 3-Dimethylamino-1-propanol
CAS 3179-63-3
Molecular Formula C5H13NO
Linear Formula (CH3)2N(CH2)3OH
Formula Weight 103.16
MDL Number MFCD00002945
UN Number UN2734
Beilstein 1732032
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA128000F

 
alfa aesar™
A128000F
2500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 103.16
Linear Formula (CH3)2N(CH2)3OH
Chemical Name or Material 3-Dimethylamino-1-propanol
TSCA TSCA
Assay 99%
Molecular Formula C5H13NO
Melting Point −35°C
Boiling Point 162° to 164°C
MDL Number MFCD00002945
CAS 3179-63-3
UN Number UN2734
Beilstein 1732032
Quantity 2500mL
Flash Point 54°C (129°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H226-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
RTECS Number UB3155000

Applications
3-Dimethylamino-1-propanol, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
It is miscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H226-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.