missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Dimethylamino-1-(3-pyridyl)-2-propen-1-one, 97%

Manufacturer:  Alfa Aesar™ H54187MD

 View more versions of this product

Catalog No. AAH54187MD



Description

Description

Applications
3-Dimethylamino-1-(3-pyridyl)-2-propen-1-one is used ads pharmaceutical intermediate, an imatinib intermediate.

Solubility
Soluble in chloroform and dichloromethane.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, acids bases.
Specifications

Specifications

3-Dimethylamino-1-(3-pyridyl)-2-propen-1-one
55314-16-4
MFCD00115180
MZLRFUCMBQWLNV-FNORWQNLSA-N
(E)-3-(dimethylamino)-1-pyridin-3-ylprop-2-en-1-one
5369165
97%
250mg
C10H12N2O
1-3-pyridyl-3-dimethylamino-2-propen-1-one
CN(C)C=CC(=O)C1=CN=CC=C1
176.219
176.22

Chemical Identifiers

55314-16-4
176.219
MZLRFUCMBQWLNV-FNORWQNLSA-N
5369165
CN(C)C=CC(=O)C1=CN=CC=C1
C10H12N2O
MFCD00115180
1-3-pyridyl-3-dimethylamino-2-propen-1-one
(E)-3-(dimethylamino)-1-pyridin-3-ylprop-2-en-1-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents