Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Dimethylamino-1,2,4-dithiazole-5-thione, 97%

$61.03 - $221.50

Chemical Identifiers

Chemical Name or Material 3-Dimethylamino-1,2,4-dithiazole-5-thione
CAS 29220-04-0
Molecular Formula C4H6N2S3
Formula Weight 178.31
MDL Number MFCD00052567
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2057403

  SDS
alfa aesar™
B2057403
1g Each for $61.03

AAB2057406

  SDS
alfa aesar™
B2057406
5g Each for $221.50
Description & Specifications

Specifications

Formula Weight 178.31
Chemical Name or Material 3-Dimethylamino-1,2,4-dithiazole-5-thione
Assay 97%
Molecular Formula C4H6N2S3
Melting Point 175° to 177°C
MDL Number MFCD00052567
CAS 29220-04-0
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
3-Dimethylamino-1,2,4-dithiazole-5-thione, is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.