missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Difluoromethyl)-1-fluorobenzene, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC451390050

 View more versions of this product

Catalog No. AC451390050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3-(Difluoromethyl)-1-fluorobenzene
74°C (130.0 Torr)
26029-52-7
C7H5F3
IKCXLEHVIIUYOA-UHFFFAOYSA-N
1-(difluoromethyl)-3-fluorobenzene
2761203
97%
23°C
5g
96% min. (GC)
1-difluoromethyl-3-fluorobenzene, 1-fluoro-3-difluoromethyl benzene, 3-difluoromethyl fluorobenzene, 3-difluoromethyl-1-fluorobenzene, benzene, 1-difluoromethyl-3-fluoro, intermediates-zcf02695
C1=CC(=CC(=C1)F)C(F)F
146.11
146.11

Chemical Identifiers

26029-52-7
146.11
1-difluoromethyl-3-fluorobenzene, 1-fluoro-3-difluoromethyl benzene, 3-difluoromethyl fluorobenzene, 3-difluoromethyl-1-fluorobenzene, benzene, 1-difluoromethyl-3-fluoro, intermediates-zcf02695
1-(difluoromethyl)-3-fluorobenzene
C7H5F3
IKCXLEHVIIUYOA-UHFFFAOYSA-N
2761203
C1=CC(=CC(=C1)F)C(F)F
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Warning

SDS Documents