missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(Difluoromethyl)-1-fluorobenzene, 97%, ACROS Organics™
Chemical Identifiers
CAS | 26029-52-7 |
---|---|
Molecular Formula | C7H5F3 |
Molecular Weight (g/mol) | 146.11 |
InChI Key | IKCXLEHVIIUYOA-UHFFFAOYSA-N |
Synonym | 1-difluoromethyl-3-fluorobenzene, 1-fluoro-3-difluoromethyl benzene, 3-difluoromethyl fluorobenzene, 3-difluoromethyl-1-fluorobenzene, benzene, 1-difluoromethyl-3-fluoro, intermediates-zcf02695 |
PubChem CID | 2761203 |
IUPAC Name | 1-(difluoromethyl)-3-fluorobenzene |
SMILES | C1=CC(=CC(=C1)F)C(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC451390010
|
Thermo Fisher Scientific
AC451390010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC451390050
|
Thermo Fisher Scientific
AC451390050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
26029-52-7 | |
146.11 | |
1-difluoromethyl-3-fluorobenzene, 1-fluoro-3-difluoromethyl benzene, 3-difluoromethyl fluorobenzene, 3-difluoromethyl-1-fluorobenzene, benzene, 1-difluoromethyl-3-fluoro, intermediates-zcf02695 | |
1-(difluoromethyl)-3-fluorobenzene |
C7H5F3 | |
IKCXLEHVIIUYOA-UHFFFAOYSA-N | |
2761203 | |
C1=CC(=CC(=C1)F)C(F)F |
Specifications
23°C | |
74°C (130.0 Torr) | |
26029-52-7 | |
C7H5F3 | |
IKCXLEHVIIUYOA-UHFFFAOYSA-N | |
1-(difluoromethyl)-3-fluorobenzene | |
2761203 | |
97% |
Glass Bottle | |
1g | |
96% min. (GC) | |
1-difluoromethyl-3-fluorobenzene, 1-fluoro-3-difluoromethyl benzene, 3-difluoromethyl fluorobenzene, 3-difluoromethyl-1-fluorobenzene, benzene, 1-difluoromethyl-3-fluoro, intermediates-zcf02695 | |
C1=CC(=CC(=C1)F)C(F)F | |
146.11 | |
146.11 | |
3-(Difluoromethyl)-1-fluorobenzene |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Warning