Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Difluoromethoxy)cinnamic acid, 97%

$299.85 - $299.85

Chemical Identifiers

Chemical Name or Material 3-(Difluoromethoxy)cinnamic acid
Molecular Formula C10H8F2O3
Formula Weight 214.17
MDL Number MFCD06205319
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3198003

  SDS
alfa aesar™
H3198003
1g Each for $299.85
Description & Specifications

Specifications

Formula Weight 214.17
Quantity 1g
MDL Number MFCD06205319
Melting Point 107° to 109°C
Assay 97%
Health Hazard 1 Warning
Chemical Name or Material 3-(Difluoromethoxy)cinnamic acid
Molecular Formula C10H8F2O3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.