Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Difluoromethoxy)benzylamine, 98%

Chemical Identifiers

Chemical Name or Material 3-(Difluoromethoxy)benzylamine
CAS 244022-71-7
Molecular Formula C8H9F2NO
Formula Weight 173.16
MDL Number MFCD00236230
UN Number UN2735
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5032203

  SDS
alfa aesar™
H5032203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5032206

  SDS
alfa aesar™
H5032206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 173.16
Health Hazard 1 Danger
MDL Number MFCD00236230
Chemical Name or Material 3-(Difluoromethoxy)benzylamine
Quantity 1g
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Recommended Storage 2° to 8°C
Assay 98%
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
UN Number UN2735
CAS 244022-71-7
Molecular Formula C8H9F2NO
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

Applications
3-(Difluoromethoxy)benzylamine is used as pharmaceutical intermediate.

Solubility
Difficult to mix.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

2° to 8°C