missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Difluoromethoxy)aniline, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC442320010

 View more versions of this product

Catalog No. AC442320010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-(Difluoromethoxy)aniline
Glass bottle
Colorless to Yellow
22236-08-4
100.0
C7H7F2NO
RSOFZRXRIPVBBM-UHFFFAOYSA-N
3-(difluoromethoxy)aniline
2774110
Liquid
Authentic
72.0°C to 76.0°C (4.0mmHg)
1g
96.0
96% min. (GC)
3-amino-alpha,alpha-difluoroanisole, 3-difluoromethoxy aniline, 3-difluoromethoxy phenylamine, 3-difluoromethoxyphenylamine, a,a-difluoro-m-anisidine, acmc-1ckmq, benzenamine, 3-difluoromethoxy, m-difluoromethoxyaniline, pubchem8466
C1=CC(=CC(=C1)OC(F)F)N
159.14
159.14
97%

Chemical Identifiers

22236-08-4
159.14
3-amino-alpha,alpha-difluoroanisole, 3-difluoromethoxy aniline, 3-difluoromethoxy phenylamine, 3-difluoromethoxyphenylamine, a,a-difluoro-m-anisidine, acmc-1ckmq, benzenamine, 3-difluoromethoxy, m-difluoromethoxyaniline, pubchem8466
3-(difluoromethoxy)aniline
C7H7F2NO
RSOFZRXRIPVBBM-UHFFFAOYSA-N
2774110
C1=CC(=CC(=C1)OC(F)F)N
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents