missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Difluoromethoxy)aniline, 97%

$69.96 - $963.22

Chemical Identifiers

CAS 22236-08-4
Molecular Formula C7H7F2NO
Molecular Weight (g/mol) 159.136
MDL Number MFCD00236221
InChI Key RSOFZRXRIPVBBM-UHFFFAOYSA-N
Synonym 3-amino-alpha,alpha-difluoroanisole
PubChem CID 2774110
IUPAC Name 3-(difluoromethoxy)aniline
SMILES C1=CC(=CC(=C1)OC(F)F)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5346803 View Documents Alfa Aesar™
H5346803
1g Each for $69.96
 
AAH5346806 View Documents Alfa Aesar™
H5346806
5g Each for $235.57
 
AAH5346814 View Documents Alfa Aesar™
H5346814
25g Each for $963.22
 
Specifications

Chemical Identifiers

22236-08-4
159.136
RSOFZRXRIPVBBM-UHFFFAOYSA-N
2774110
C1=CC(=CC(=C1)OC(F)F)N
C7H7F2NO
MFCD00236221
3-amino-alpha,alpha-difluoroanisole
3-(difluoromethoxy)aniline

Specifications

1.276
Air Sensitive
1g
C7H7F2NO
3-amino-alpha,alpha-difluoroanisole
C1=CC(=CC(=C1)OC(F)F)N
159.136
159.14
3-(Difluoromethoxy)aniline
1.51
201°C to 202°C
22236-08-4
MFCD00236221
RSOFZRXRIPVBBM-UHFFFAOYSA-N
3-(difluoromethoxy)aniline
2774110
97%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures