missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Difluoromethoxy)aniline, 97%

Manufacturer:  Alfa Aesar™ H5346803

 View more versions of this product

Catalog No. AAH5346803



Specifications

Specifications

3-(Difluoromethoxy)aniline
1.51
201°C to 202°C
22236-08-4
MFCD00236221
RSOFZRXRIPVBBM-UHFFFAOYSA-N
3-(difluoromethoxy)aniline
2774110
97%
1.276
Air Sensitive
1g
C7H7F2NO
3-amino-alpha,alpha-difluoroanisole, 3-difluoromethoxy aniline, 3-difluoromethoxy phenylamine, 3-difluoromethoxyphenylamine, a,a-difluoro-m-anisidine, acmc-1ckmq, benzenamine, 3-difluoromethoxy, m-difluoromethoxyaniline, pubchem8466
C1=CC(=CC(=C1)OC(F)F)N
159.136
159.14

Chemical Identifiers

22236-08-4
159.136
RSOFZRXRIPVBBM-UHFFFAOYSA-N
2774110
C1=CC(=CC(=C1)OC(F)F)N
C7H7F2NO
MFCD00236221
3-amino-alpha,alpha-difluoroanisole, 3-difluoromethoxy aniline, 3-difluoromethoxy phenylamine, 3-difluoromethoxyphenylamine, a,a-difluoro-m-anisidine, acmc-1ckmq, benzenamine, 3-difluoromethoxy, m-difluoromethoxyaniline, pubchem8466
3-(difluoromethoxy)aniline
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents