Please login to your online account to display your discounted pricing

3-Diethylaminophenol, Granulated 99%, ACROS Organics™

$66.06 - $66.06

Chemical Identifiers

Chemical Name or Material 3-Diethylaminophenol
Name Note 99%
CAS 91-68-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H15NO
Linear Formula (C2H5)2NC6H4OH
Formula Weight 165.23
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC146631000

  SDS
thermo fisher scientific
146631000
Glass bottle 100g Each for $66.06
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Flakes or Granules
Formula Weight 165.23
Quantity 100g
Color Gray-brown to rose or red
EINECS Number 202-090-9
TSCA TSCA
Melting Point 70° to 73°C
RTECS Number SL0550500
Beilstein 13, 408
CAS Max % 100.0
Packaging Glass bottle
Synonym N, N-Diethyl-3-aminophenol
CAS Min % 98.5
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face, hands and any exposed skin thoroughly after handling.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not eat, drink or smoke when using this product.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3-Diethylaminophenol
Name Note 99%
Assay Percent Range 99%
Molecular Formula C10H15NO
Linear Formula (C2H5)2NC6H4OH
MDL Number MFCD00002265
CAS 91-68-9
Health Hazard 2 GHS P Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Safety and Handling

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face, hands and any exposed skin thoroughly after handling.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not eat, drink or smoke when using this product.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.