missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Diethylamino-1-propyne, 98+%

Manufacturer:  Alfa Aesar™ L1223714

 View more versions of this product

Catalog No. AAL1223714



Description

Description

Applications
3-Diethylamino-1-propyne is used as functional leveling agent, brightener.

Solubility
Soluble in acid. Insoluble in water.

Notes
Incompatible with heat, light, carbondioxide, heavy metal powders. Air & Light Sensitive. Store in the dark. Store under dry inert gas.
Specifications

Specifications

3-Diethylamino-1-propyne
Amine-like
25g
C7H13N
MFCD00039881
1098468
JZJXKEWVUBVOEH-UHFFFAOYSA-N
N,N-diethylprop-2-yn-1-amine
20010
≥98%
20°C (68°F)
119°C to 120°C
4079-68-9
(CH3CH2)2NCH2C≡CH
UN2733
1-diethylamino-2-propyne, diethylpropargylamine, 2-propyn-1-amine, n,n-diethyl, n,n-diethylpropargylamine, n,n-diethyl-2-propynylamine, 3-diethylamino propyne, 3-diethylamino-1-propyne, 3-n,n-diethylamino propyne, 1-n,n-diethylamino-2-propyne, 2-propynylamine, n,n-diethyl
CCN(CC)CC#C
111.188
111.19

Chemical Identifiers

4079-68-9
111.188
JZJXKEWVUBVOEH-UHFFFAOYSA-N
20010
CCN(CC)CC#C
C7H13N
MFCD00039881
1-diethylamino-2-propyne, diethylpropargylamine, 2-propyn-1-amine, n,n-diethyl, n,n-diethylpropargylamine, n,n-diethyl-2-propynylamine, 3-diethylamino propyne, 3-diethylamino-1-propyne, 3-n,n-diethylamino propyne, 1-n,n-diethylamino-2-propyne, 2-propynylamine, n,n-diethyl
N,N-diethylprop-2-yn-1-amine
Safety and Handling

Safety and Handling

GHS H Statement
H225-H311-H314-H318-H302-H332
Highly flammable liquid and vapor.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P210-P280-P305+P351+P338-P309-P310
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

RTECSNumber : UK5775000

TSCA : TSCA

Documents