missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Diethylamino-1-propyl isothiocyanate, 97%

Manufacturer:  Alfa Aesar™ L1075003

 View more versions of this product

Catalog No. AAL1075003



Specifications

Specifications

3-Diethylamino-1-propyl isothiocyanate
Irritating
1g
C8H16N2S
UN2922
3-diethylamino propyl isothiocyanate, diethyl 3-isothiocyanatopropyl amine, 3-diethylamino propylisothiocyanate, diethyl-3-isothiocyanato-propyl-amine, acmc-20aniz, n,n-diethyl-3-isothiocyanato-propan-1-amine, 3-diethylaminopropyl isothiocyanate, 3-diethylamino propanisothiocyanate, 3-diethylamino-1-propyl isothiocyanate, 3-n,n-diethylamino propylisothiocyanate
CCN(CC)CCCN=C=S
172.29
172.29
>110°C (230°F)
94°C to 95°C (3.5mmHg)
2626-52-0
MFCD00041133
1757446
QPBVKSLJBRCKIF-UHFFFAOYSA-N
N,N-diethyl-3-isothiocyanatopropan-1-amine
2758240
97%

Chemical Identifiers

2626-52-0
172.29
QPBVKSLJBRCKIF-UHFFFAOYSA-N
2758240
CCN(CC)CCCN=C=S
C8H16N2S
MFCD00041133
3-diethylamino propyl isothiocyanate, diethyl 3-isothiocyanatopropyl amine, 3-diethylamino propylisothiocyanate, diethyl-3-isothiocyanato-propyl-amine, acmc-20aniz, n,n-diethyl-3-isothiocyanato-propan-1-amine, 3-diethylaminopropyl isothiocyanate, 3-diethylamino propanisothiocyanate, 3-diethylamino-1-propyl isothiocyanate, 3-n,n-diethylamino propylisothiocyanate
N,N-diethyl-3-isothiocyanatopropan-1-amine
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents