Please login to your online account to display your discounted pricing

3-(Diethylamino)-1,2-propanediol, 98%, ACROS Organics™


Quantity 250mL

Manufacturer: thermo fisher scientific  198502500

 View more versions of this product

Catalog No. AC198502500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Viscous Liquid
Formula Weight 147.22
Assay ≥97.5%
Quantity 250mL
Color Clear colorless to yellow
TSCA TSCA
EINECS Number 210-693-3
Density 96%g/mL
Beilstein 04, 302
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 1 GHS Signal Word: Danger
CAS Min % 97.5
Refractive Index 1.4592 to 1.4612 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 621-56-7
Linear Formula (C2H5)2NCH2CH(OH)CH2OH
Molecular Formula C7H17NO2
Chemical Name or Material 3-(Diethylamino)-1, 2-propanediol
Name Note 98%
Assay Percent Range 98%
MDL Number MFCD00004716

Specifications

Chemical Name or Material 3-(Diethylamino)-1, 2-propanediol
Name Note 98%
CAS 621-56-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H17NO2
Linear Formula (C2H5)2NCH2CH(OH)CH2OH
Formula Weight 147.22
MDL Number MFCD00004716
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents