Please login to your online account to display your discounted pricing

3-(Diethylamino)-1,2-propanediol, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-(Diethylamino)-1, 2-propanediol
Name Note 98%
CAS 621-56-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H17NO2
Linear Formula (C2H5)2NCH2CH(OH)CH2OH
Formula Weight 147.22
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC198502500

  SDS
thermo fisher scientific
198502500
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Viscous Liquid
Assay ≥97.5%
Formula Weight 147.22
Quantity 250mL
Color Clear colorless to yellow
EINECS Number 210-693-3
TSCA TSCA
Density 96%g/mL
Beilstein 04, 302
CAS Max % 100.0
Chemical Name or Material 3-(Diethylamino)-1, 2-propanediol
Refractive Index 1.4592 to 1.4612 (20°C, 589nm)
Name Note 98%
Assay Percent Range 98%
Molecular Formula C7H17NO2
Linear Formula (C2H5)2NCH2CH(OH)CH2OH
MDL Number MFCD00004716
CAS 621-56-7
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.