missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Di-tert-butylphosphino)propylamine, 10% w/w soln. in THF

$31.61 - $431.80

Chemical Identifiers

CAS 1196147-72-4
Molecular Formula C11H26NP
Molecular Weight (g/mol) 203.31
MDL Number MFCD17013990
InChI Key PREMCOLELPAPBB-UHFFFAOYSA-N
Synonym 3-di-t-butylphosphino propylamine, 3-aminopropyldi-tert-butylphosphine, 3-di-tert-butylphosphino propylamine, 3-aminopropyl di-tert-butyl phosphane, 3-aminopropyl di-tert-butyl phosphane
PubChem CID 72376443
IUPAC Name 3-ditert-butylphosphanylpropan-1-amine
SMILES CC(C)(C)P(CCCN)C(C)(C)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6094303 View Documents Alfa Aesar™
H6094303
1g Each for $31.61
 
AAH6094306 View Documents Alfa Aesar™
H6094306
5g Each for $112.58
 
AAH6094314 View Documents Alfa Aesar™
H6094314
25g Each for $431.80
 
Specifications

Chemical Identifiers

1196147-72-4
203.31
PREMCOLELPAPBB-UHFFFAOYSA-N
72376443
CC(C)(C)P(CCCN)C(C)(C)C
C11H26NP
MFCD17013990
3-di-t-butylphosphino propylamine, 3-aminopropyldi-tert-butylphosphine, 3-di-tert-butylphosphino propylamine, 3-aminopropyl di-tert-butyl phosphane, 3-aminopropyl di-tert-butyl phosphane
3-ditert-butylphosphanylpropan-1-amine

Specifications

1g
C11H26NP
MFCD17013990
3-di-t-butylphosphino propylamine, 3-aminopropyldi-tert-butylphosphine, 3-di-tert-butylphosphino propylamine, 3-aminopropyl di-tert-butyl phosphane, 3-aminopropyl di-tert-butyl phosphane
PREMCOLELPAPBB-UHFFFAOYSA-N
3-ditert-butylphosphanylpropan-1-amine
72376443
3-(Di-tert-butylphosphino)propylamine
1196147-72-4
[(CH3)3C]2PCH2CH2CH2NH2
UN2733
10% w/w soln. in THF
CC(C)(C)P(CCCN)C(C)(C)C
203.31
203.3
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H351-H335
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.