missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Di-n-propylsulfamoyl)benzeneboronic acid, 97%

$188.04 - $601.09

Chemical Identifiers

CAS 1449145-31-6
Molecular Formula C12H20BNO4S
Molecular Weight (g/mol) 285.165
MDL Number MFCD20265249
InChI Key NUYSZOMJDYKRCE-UHFFFAOYSA-N
Synonym 3-di-n-propylsulfamoyl benzeneboronic acid
PubChem CID 73995769
IUPAC Name [3-(dipropylsulfamoyl)phenyl]boronic acid
SMILES B(C1=CC(=CC=C1)S(=O)(=O)N(CCC)CCC)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54354MD View Documents Alfa Aesar™
H54354MD
250mg Each for $188.04
 
AAH5435403 View Documents Alfa Aesar™
H5435403
1g Each for $601.09
 
Specifications

Chemical Identifiers

1449145-31-6
285.165
NUYSZOMJDYKRCE-UHFFFAOYSA-N
73995769
B(C1=CC(=CC=C1)S(=O)(=O)N(CCC)CCC)(O)O
C12H20BNO4S
MFCD20265249
3-di-n-propylsulfamoyl benzeneboronic acid
[3-(dipropylsulfamoyl)phenyl]boronic acid

Specifications

250mg
C12H20BNO4S
3-di-n-propylsulfamoyl benzeneboronic acid
B(C1=CC(=CC=C1)S(=O)(=O)N(CCC)CCC)(O)O
285.165
285.17
3-(Di-n-propylsulfamoyl)benzeneboronic acid
1449145-31-6
MFCD20265249
NUYSZOMJDYKRCE-UHFFFAOYSA-N
[3-(dipropylsulfamoyl)phenyl]boronic acid
73995769
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures