missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Di-n-propylsulfamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H54354MD

 View more versions of this product

Catalog No. AAH54354MD



Specifications

Specifications

3-(Di-n-propylsulfamoyl)benzeneboronic acid
1449145-31-6
MFCD20265249
NUYSZOMJDYKRCE-UHFFFAOYSA-N
[3-(dipropylsulfamoyl)phenyl]boronic acid
73995769
97%
250mg
C12H20BNO4S
3-di-n-propylsulfamoyl benzeneboronic acid
B(C1=CC(=CC=C1)S(=O)(=O)N(CCC)CCC)(O)O
285.165
285.17

Chemical Identifiers

1449145-31-6
285.165
NUYSZOMJDYKRCE-UHFFFAOYSA-N
73995769
B(C1=CC(=CC=C1)S(=O)(=O)N(CCC)CCC)(O)O
C12H20BNO4S
MFCD20265249
3-di-n-propylsulfamoyl benzeneboronic acid
[3-(dipropylsulfamoyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents