Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Di-n-propylamino-1-propyne, 98%

Chemical Identifiers

Chemical Name or Material 3-Di-n-propylamino-1-propyne
CAS 6323-79-1
Molecular Formula C9H17N
Formula Weight 139.24
MDL Number MFCD00041626
UN Number UN2733
Beilstein 1746300
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1163306

  SDS
alfa aesar™
L1163306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1163303

  SDS
alfa aesar™
L1163303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 139.24
Chemical Name or Material 3-Di-n-propylamino-1-propyne
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Molecular Formula C9H17N
UN Number UN2733
CAS 6323-79-1
Quantity 5g
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Boiling Point 157° to 159°C
Assay 98%
MDL Number MFCD00041626
Flash Point 55°C (131°F)
Odor Amine-like
Beilstein 1746300
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.