missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Di-n-propylamino-1-propyne, 98%

Manufacturer:  Alfa Aesar™ L1163306

 View more versions of this product

Catalog No. AAL1163306



Specifications

Specifications

3-Di-n-propylamino-1-propyne
Amine-like
5g
C9H17N
UN2733
3-di-n-propylamino-1-propyne, n,n-dipropyl-2-propyn-1-amine, propargyldipropylamine, n,n-dipropylprop-2-yn-1-amine, 3-dipropylamino-1-propyne, 2-propyn-1-amine, n,n-dipropyl, prop-2-ynyldipropylamine, dipropylpropargylamine, propargyl di-n-propyl amine, n,n-dipropyl-2-propynylamine
CCCN(CCC)CC#C
139.242
139.24
55°C (131°F)
157°C to 159°C
6323-79-1
MFCD00041626
1746300
ATTMKPWGJILFFV-UHFFFAOYSA-N
N-propyl-N-prop-2-ynylpropan-1-amine
138724
98%

Chemical Identifiers

6323-79-1
139.242
ATTMKPWGJILFFV-UHFFFAOYSA-N
138724
CCCN(CCC)CC#C
C9H17N
MFCD00041626
3-di-n-propylamino-1-propyne, n,n-dipropyl-2-propyn-1-amine, propargyldipropylamine, n,n-dipropylprop-2-yn-1-amine, 3-dipropylamino-1-propyne, 2-propyn-1-amine, n,n-dipropyl, prop-2-ynyldipropylamine, dipropylpropargylamine, propargyl di-n-propyl amine, n,n-dipropyl-2-propynylamine
N-propyl-N-prop-2-ynylpropan-1-amine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

Documents