Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Di-n-butylamino)-1-propyne, 98%

$141.47 - $141.47

Chemical Identifiers

Chemical Name or Material 3-(Di-n-butylamino)-1-propyne
CAS 6336-58-9
Molecular Formula C11H21N
Formula Weight 167.3
MDL Number MFCD00041673
Beilstein 1752173
Synonym N,N-Di-n-butylpropargylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1084406

  SDS
alfa aesar™
L1084406
5g Each for $141.47
Description & Specifications

Specifications

Formula Weight 167.3
Chemical Name or Material 3-(Di-n-butylamino)-1-propyne
Assay 98%
MDL Number MFCD00041673
CAS 6336-58-9
Beilstein 1752173
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Boiling Point 87° to 89°C (19mmHg)
Quantity 5g
Odor Amine-like
Synonym N,N-Di-n-butylpropargylamine
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Molecular Formula C11H21N
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.