missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Di-n-butylamino)-1-propyne, 98%

Manufacturer:  Alfa Aesar™ L1084403

 View more versions of this product

Catalog No. AAL1084403



Specifications

Specifications

3-(Di-n-butylamino)-1-propyne
87°C to 89°C (19mmHg)
6336-58-9
MFCD00041673
3-dibutylamino-1-propyne
CCCCN(CCCC)CC#C
167.296
167.3
Amine-like
1g
C11H21N
1752173
XTEBRELWTGWYDE-UHFFFAOYSA-N
N-butyl-N-prop-2-ynylbutan-1-amine
138732
98%

Chemical Identifiers

6336-58-9
167.296
XTEBRELWTGWYDE-UHFFFAOYSA-N
138732
CCCCN(CCCC)CC#C
C11H21N
MFCD00041673
3-dibutylamino-1-propyne
N-butyl-N-prop-2-ynylbutan-1-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Documents