Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Decyne, 98+%

Chemical Identifiers

Chemical Name or Material 3-Decyne
CAS 2384-85-2
Molecular Formula CH3(CH2)5C≡CCH2CH3
Formula Weight 138.26
MDL Number MFCD00039987
UN Number UN3295
Beilstein 1698698
Synonym Ethyl n-hexyl acetylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2026803

 
alfa aesar™
B2026803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2026806

 
alfa aesar™
B2026806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2026814

 
alfa aesar™
B2026814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 138.26
Chemical Name or Material 3-Decyne
TSCA TSCA
Assay ≥98%
Molecular Formula CH3(CH2)5C≡CCH2CH3
MDL Number MFCD00039987
CAS 2384-85-2
UN Number UN3295
Beilstein 1698698
Synonym Ethyl n-hexyl acetylene
Odor Petrol-like
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 175° to 176°C
Flash Point 56°C (132°F)
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.