Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Decen-2-one, 97%

$166.24 - $166.24

Chemical Identifiers

Chemical Name or Material 3-Decen-2-one
CAS 10519-33-2
Molecular Formula CH3(CH2)5CH=CHCOCH3
Formula Weight 154.25
MDL Number MFCD00015700
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1925614

  SDS
alfa aesar™
A1925614
25g Each for $166.24
Description & Specifications

Specifications

Formula Weight 154.25
Chemical Name or Material 3-Decen-2-one
TSCA TSCA
Assay 97%
Molecular Formula CH3(CH2)5CH=CHCOCH3
MDL Number MFCD00015700
CAS 10519-33-2
Quantity 25g