Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Decanone, 99%

$133.46 - $443.31

Chemical Identifiers

Chemical Name or Material 3-Decanone
CAS 928-80-3
Molecular Formula CH3(CH2)6COCH2CH3
Formula Weight 156.27
MDL Number MFCD00009554
Beilstein 1748021
Synonym Ethyl n-heptyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2090914

  SDS
alfa aesar™
B2090914
25g Each for $133.46

AAB2090922

  SDS
alfa aesar™
B2090922
100g Each for $443.31
Description & Specifications

Specifications

Formula Weight 156.27
Boiling Point 204° to 205°C
MDL Number MFCD00009554
CAS 928-80-3
Melting Point −4°C
Quantity 25g
Flash Point 81°C (178°F)
Synonym Ethyl n-heptyl ketone
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 3-Decanone
TSCA TSCA
Assay 99%
Molecular Formula CH3(CH2)6COCH2CH3
Beilstein 1748021
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.