Please login to your online account to display your discounted pricing

3-Cyclopropylpropargyl diethyl acetal 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Cyclopropylpropargyl diethyl acetal
CAS 146195-69-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 97%
Molecular Formula C10H16O2
Linear Formula C3H5CCCH(OCH2CH3)2
Formula Weight 168.24
MDL Number MFCD12912455
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC391650010

  SDS
thermo fisher scientific
AC391650010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC391650050

  SDS
thermo fisher scientific
AC391650050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 168.24
Quantity 1g
Color Clear yellow
Density 0.97g/mL
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Refractive Index 1.4520 to 1.4540 (20°C, 589nm)
Molecular Formula C10H16O2
Linear Formula C3H5CCCH(OCH2CH3)2
MDL Number MFCD12912455
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Chemical Name or Material 3-Cyclopropylpropargyl diethyl acetal
Infrared Spectrum Authentic
Assay Percent Range 97%
CAS 146195-69-9
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.