Learn More
3-Cyclopropylpropargyl diethyl acetal, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 391650010
Specifications
3-Cyclopropylpropargyl diethyl acetal | |
96.0 | |
0.9700g/mL | |
83°C | |
97.5% min. (GC) | |
C10H16O2 | |
C3H5CCCH(OCH2CH3)2 | |
1g | |
3-cyclopropylpropiolaldehyde diethyl acetal, 3,3-diethoxyprop-1-yn-1-yl cyclopropane, 3-cyclopropylpropargyl diethyl acetal, 3-cyclopropylpropargyl aldehyde diethyl acetal | |
CCOC(C#CC1CC1)OCC | |
168.24 | |
168.24 |
146195-69-9 | |
100.0 | |
236°C | |
Authentic | |
Glass bottle | |
1.4520 to 1.4540 (20°C, 589nm) | |
MFCD12912455 | |
0.97 | |
WOSJTIAXBXVXLF-UHFFFAOYSA-N | |
3,3-diethoxyprop-1-ynylcyclopropane | |
42580279 | |
97% |
Chemical Identifiers
146195-69-9 | |
168.24 | |
WOSJTIAXBXVXLF-UHFFFAOYSA-N | |
42580279 | |
CCOC(C#CC1CC1)OCC |
C10H16O2 | |
MFCD12912455 | |
3-cyclopropylpropiolaldehyde diethyl acetal, 3,3-diethoxyprop-1-yn-1-yl cyclopropane, 3-cyclopropylpropargyl diethyl acetal, 3-cyclopropylpropargyl aldehyde diethyl acetal | |
3,3-diethoxyprop-1-ynylcyclopropane |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO â Research Use Only