missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Cyclopropylmethoxy-4-difluoromethoxybenzoic acid, 98%

Manufacturer:  Alfa Aesar™ H64098MD

 View more versions of this product

Catalog No. AAH64098MD



Specifications

Specifications

3-Cyclopropylmethoxy-4-difluoromethoxybenzoic acid
250mg
C12H12F2O4
11413145
IGFDIFLMMLWKKY-UHFFFAOYSA-N
3-(cyclopropylmethoxy)-4-(difluoromethoxy)benzoic acid
2394006
Solid
118°C to 120°C
162401-62-9
MFCD04621687
3-cyclopropylmethoxy-4-difluoromethoxy benzoic acid, 3-cyclopropylmethoxy-4-difluoromethoxy-benzoic acid, benzoic acid, 3-cyclopropylmethoxy-4-difluoromethoxy, roflumilast intermediate, roflumilast intermediate b, 3-cyclopropylmethoxy-4-difluoromethoxybenzoicacid, 3-cyclopropylmethoxy-4-difluoromethoxy-benzoic aci, 3-cyclopropylmethoxy 4-difluoromethoxy benzoic acid, 3-cyclopropylmethoxy-4-difluoromethoxy-benzoic acid
C1CC1COC2=C(C=CC(=C2)C(=O)O)OC(F)F
258.221
258.22
98%

Chemical Identifiers

162401-62-9
258.221
IGFDIFLMMLWKKY-UHFFFAOYSA-N
2394006
C1CC1COC2=C(C=CC(=C2)C(=O)O)OC(F)F
C12H12F2O4
MFCD04621687
3-cyclopropylmethoxy-4-difluoromethoxy benzoic acid, 3-cyclopropylmethoxy-4-difluoromethoxy-benzoic acid, benzoic acid, 3-cyclopropylmethoxy-4-difluoromethoxy, roflumilast intermediate, roflumilast intermediate b, 3-cyclopropylmethoxy-4-difluoromethoxybenzoicacid, 3-cyclopropylmethoxy-4-difluoromethoxy-benzoic aci, 3-cyclopropylmethoxy 4-difluoromethoxy benzoic acid, 3-cyclopropylmethoxy-4-difluoromethoxy-benzoic acid
3-(cyclopropylmethoxy)-4-(difluoromethoxy)benzoic acid
SDS Documents