missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Cyclopropyl-1H-pyrazole, 98%

Manufacturer:  Alfa Aesar™ H64069MD

 View more versions of this product

Catalog No. AAH64069MD



Specifications

Specifications

3-Cyclopropyl-1H-pyrazole
250mg
C6H8N2
1561659
TXWDVWSJMDFNQY-UHFFFAOYSA-N
5-cyclopropyl-1H-pyrazole
7017816
Liquid
Colorless
100114-57-6
MFCD04970938
3-cyclopropyl-1h-pyrazole, 5-cyclopropylpyrazole, 1h-pyrazole, 3-cyclopropyl, 3-cyclopropylpyrazole, 1h-pyrazole,3-cyclopropyl, 3-cyclopropyl-2h-pyrazole, 3-cyclopropyl-1h-pyrazole 97+%, acmc-2097ms, 1h-pyrazole, 5-cyclopropyl
C1CC1C2=CC=NN2
108.144
108.14
98%

Chemical Identifiers

100114-57-6
108.144
TXWDVWSJMDFNQY-UHFFFAOYSA-N
7017816
C1CC1C2=CC=NN2
C6H8N2
MFCD04970938
3-cyclopropyl-1h-pyrazole, 5-cyclopropylpyrazole, 1h-pyrazole, 3-cyclopropyl, 3-cyclopropylpyrazole, 1h-pyrazole,3-cyclopropyl, 3-cyclopropyl-2h-pyrazole, 3-cyclopropyl-1h-pyrazole 97+%, acmc-2097ms, 1h-pyrazole, 5-cyclopropyl
5-cyclopropyl-1H-pyrazole
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents