missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Cyclopentyloxy-4-methoxybenzyl alcohol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H6244103

 View more versions of this product

Catalog No. AAH6244103


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-Cyclopentyloxy-4-methoxybenzyl alcohol
C13H18O3
1 g
JIRHAGAOHOYLNO-UHFFFAOYSA-N
(3-cyclopentyloxy-4-methoxyphenyl)methanol
11138706
97%
133332-49-7
MFCD03844627
3-cyclopentyloxy-4-methoxybenzyl alcohol, 3-cyclopentyloxy-4-methoxyphenyl methanol, 3-cyclopentyloxy-4-methoxybenzylalcohol, acmc-1cioj, 3-cyclopentoxy-4-methoxybenzyl alcohol, 3-cyclopentyloxy-4-methoxy-benzyl alcohol
COC1=C(C=C(C=C1)CO)OC2CCCC2
222.284
222.28

Chemical Identifiers

133332-49-7
222.284
JIRHAGAOHOYLNO-UHFFFAOYSA-N
11138706
COC1=C(C=C(C=C1)CO)OC2CCCC2
C13H18O3
MFCD03844627
3-cyclopentyloxy-4-methoxybenzyl alcohol, 3-cyclopentyloxy-4-methoxyphenyl methanol, 3-cyclopentyloxy-4-methoxybenzylalcohol, acmc-1cioj, 3-cyclopentoxy-4-methoxybenzyl alcohol, 3-cyclopentyloxy-4-methoxy-benzyl alcohol
(3-cyclopentyloxy-4-methoxyphenyl)methanol
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions