Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Cyclopentyl-1-propyne, 98+%

Chemical Identifiers

Chemical Name or Material 3-Cyclopentyl-1-propyne
CAS 116279-08-4
Molecular Formula C8H12
Formula Weight 108.18
MDL Number MFCD00041559
UN Number UN3295
Beilstein 2424582
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1151406

 
alfa aesar™
L1151406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1151403

 
alfa aesar™
L1151403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 108.18
Assay ≥98%
Molecular Formula C8H12
MDL Number MFCD00041559
CAS 116279-08-4
Health Hazard 2 GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Health Hazard 3 GHS P Statement
P210-P261-P301+P330+P331-P315
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Get immediate medical advice/attention.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Chemical Name or Material 3-Cyclopentyl-1-propyne
UN Number UN3295
Beilstein 2424582
Health Hazard 1 Danger
Flash Point 21°C (69°F)
Boiling Point 132° to 134°C
Quantity 5g

Applications
3-Cyclopentyl-1-propyne is an important raw material and intermediate used in organic synthesis and pharmaceuticals.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. Incompatible with strong oxidizing agents. Store at room temperature.

Safety and Handling

Danger

GHS P Statement
P210-P261-P301+P330+P331-P315
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Get immediate medical advice/attention.

GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.