missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Cyclopentyl-1-propyne, 98+%

Manufacturer:  Alfa Aesar™ L1151403

 View more versions of this product

Catalog No. AAL1151403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Applications
3-Cyclopentyl-1-propyne is an important raw material and intermediate used in organic synthesis and pharmaceuticals.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. Incompatible with strong oxidizing agents. Store at room temperature.
Specifications

Specifications

3-Cyclopentyl-1-propyne
132°C to 134°C
116279-08-4
MFCD00041559
2424582
VBTCAHMANZTRMG-UHFFFAOYSA-N
prop-2-ynylcyclopentane
521007
≥98%
21°C (69°F)
1g
C8H12
UN3295
3-cyclopentyl-1-propyne, cyclopentylpropyne, 3-cyclopentylpropyne, 2-propynylcyclopentane, 3-cyclopentylprop-1-yne, 2-propynylcyclopentane #, prop-2-ynyl-cyclopentane, acmc-1c2wa, prop-2-yn-1-ylcyclopentane, cyclopentane,2-propyn-1-yl
C#CCC1CCCC1
108.184
108.18

Chemical Identifiers

116279-08-4
108.184
VBTCAHMANZTRMG-UHFFFAOYSA-N
521007
C#CCC1CCCC1
C8H12
MFCD00041559
3-cyclopentyl-1-propyne, cyclopentylpropyne, 3-cyclopentylpropyne, 2-propynylcyclopentane, 3-cyclopentylprop-1-yne, 2-propynylcyclopentane #, prop-2-ynyl-cyclopentane, acmc-1c2wa, prop-2-yn-1-ylcyclopentane, cyclopentane,2-propyn-1-yl
prop-2-ynylcyclopentane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.

GHS P Statement
P210-P261-P301+P330+P331-P315
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Get immediate medical advice/attention.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

Documents