missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Cyclopentene-1-carboxylic acid, 95%, Thermo Scientific™

$177.36 - $177.36

Chemical Identifiers

CAS 7686-77-3
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00101764
InChI Key XVSYDLITVYBCBD-UHFFFAOYSA-N
Synonym 3-cyclopentene-1-carboxylic acid, cyclopent-3-enecarboxylic acid, 3-cyclopentenecarboxylic acid, 3-cyclopentene-1-carhoxylic acid, cyclopent-3-ene carboxylic acid, cyclopentene-3-carboxylic acid, ghl.pd_mitscher_leg0.243, 3-cyclopentene-1-carboxylicacid, acmc-1bh22, ksc377a8b
PubChem CID 853756
IUPAC Name cyclopent-3-ene-1-carboxylic acid
SMILES C1C=CCC1C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2545603
View Documents Promotion Details
Thermo Scientific Chemicals
B2545603
1 g
Each for $177.36
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Cyclopentene-1-carboxylic acid is used as an intermediate in the production of dolasetron mesylate.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Specifications

Chemical Identifiers

7686-77-3
112.128
XVSYDLITVYBCBD-UHFFFAOYSA-N
853756
C1C=CCC1C(=O)O
C6H8O2
MFCD00101764
3-cyclopentene-1-carboxylic acid, cyclopent-3-enecarboxylic acid, 3-cyclopentenecarboxylic acid, 3-cyclopentene-1-carhoxylic acid, cyclopent-3-ene carboxylic acid, cyclopentene-3-carboxylic acid, ghl.pd_mitscher_leg0.243, 3-cyclopentene-1-carboxylicacid, acmc-1bh22, ksc377a8b
cyclopent-3-ene-1-carboxylic acid

Specifications

7686-77-3
215°C
C6H8O2
MFCD00101764
3-cyclopentene-1-carboxylic acid, cyclopent-3-enecarboxylic acid, 3-cyclopentenecarboxylic acid, 3-cyclopentene-1-carhoxylic acid, cyclopent-3-ene carboxylic acid, cyclopentene-3-carboxylic acid, ghl.pd_mitscher_leg0.243, 3-cyclopentene-1-carboxylicacid, acmc-1bh22, ksc377a8b
C1C=CCC1C(=O)O
112.128
112.13
3-Cyclopentene-1-carboxylic acid
1.084
>110°C (230°F)
1.469
1 g
XVSYDLITVYBCBD-UHFFFAOYSA-N
cyclopent-3-ene-1-carboxylic acid
853756
95%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'