missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Cyclohexylmethylsulfamoyl)benzeneboronic acid, 97%

$211.27 - $675.45

Chemical Identifiers

CAS 848779-90-8
Molecular Formula C13H20BNO4S
Molecular Weight (g/mol) 297.176
MDL Number MFCD20265269
InChI Key JDTPYMILBXMOQE-UHFFFAOYSA-N
Synonym 3-cyclohexylmethylsulfamoyl benzeneboronic acid
PubChem CID 73995807
IUPAC Name [3-(cyclohexylmethylsulfamoyl)phenyl]boronic acid
SMILES B(C1=CC(=CC=C1)S(=O)(=O)NCC2CCCCC2)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54431MD View Documents Alfa Aesar™
H54431MD
250mg Each for $211.27
 
AAH5443103 View Documents Alfa Aesar™
H5443103
1g Each for $675.45
 
Specifications

Chemical Identifiers

848779-90-8
297.176
JDTPYMILBXMOQE-UHFFFAOYSA-N
73995807
B(C1=CC(=CC=C1)S(=O)(=O)NCC2CCCCC2)(O)O
C13H20BNO4S
MFCD20265269
3-cyclohexylmethylsulfamoyl benzeneboronic acid
[3-(cyclohexylmethylsulfamoyl)phenyl]boronic acid

Specifications

250mg
C13H20BNO4S
3-cyclohexylmethylsulfamoyl benzeneboronic acid
B(C1=CC(=CC=C1)S(=O)(=O)NCC2CCCCC2)(O)O
297.176
297.18
3-(Cyclohexylmethylsulfamoyl)benzeneboronic acid
848779-90-8
MFCD20265269
JDTPYMILBXMOQE-UHFFFAOYSA-N
[3-(cyclohexylmethylsulfamoyl)phenyl]boronic acid
73995807
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning