missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Cyclohexylhydantoin, 95%

Manufacturer:  Alfa Aesar™ H6304806

 View more versions of this product

Catalog No. AAH6304806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Cyclohexylhydantoin
5g
95%
MFCD12137796
SLCFSZHXCYYXLK-UHFFFAOYSA-N
3-cyclohexylimidazolidine-2,4-dione
12434609
Solid
White
67518-23-4
C9H14N2O2
3-Cyclohexyl-2,4-imidazolidinedione
C1CCC(CC1)N2C(=O)CNC2=O
182.223
182.22

Chemical Identifiers

67518-23-4
182.223
SLCFSZHXCYYXLK-UHFFFAOYSA-N
12434609
C9H14N2O2
MFCD12137796
3-Cyclohexyl-2,4-imidazolidinedione
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents