missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Cyclohexyl-1-propyne, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2050506

 View more versions of this product

Catalog No. AAB2050506



Description

Description

3-Cyclohexyl-1-propyne, is used as a pharmaceutical intermediate. It can be used to produce (5-bromo-4,4,5,5-tetrafluoro-pent-2-enyl)-cyclohexane at the temperature of 40°C. It will need reagents (NH4)2S2O8, HCO2Na·2H2O and solvent dimethylformamide with the reaction time of 3 hours. It is also used as organic synthesis intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Cyclohexyl-1-propyne, is used as a pharmaceutical intermediate. It can be used to produce (5-bromo-4,4,5,5-tetrafluoro-pent-2-enyl)-cyclohexane at the temperature of 40°C. It will need reagents (NH4)2S2O8, HCO2Na·2H2O and solvent dimethylformamide with the reaction time of 3 hours. It is also used as organic synthesis intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

3-Cyclohexyl-1-propyne
0.845
35°C (95°F)
1.462
5 g
1849402
Slightly soluble in water.
C#CCC1CCCCC1
122.211
122.21
17715-00-3
157°C to 158°C
C9H14
MFCD00041562
UN3295
3-cyclohexyl-1-propyne, prop-2-yn-1-ylcyclohexane, 1-prop-2-ynylcyclohexane, 3-cyclohexylpropyne, 2-propynylcyclohexane, propyne, 3-cyclohexyl, cyclohexane, 2-propynyl-9ci, prop-2-ynyl-cyclohexane, cyclohexane, 2-propynyl, 3-cyclohexylprop-1-yne
UARFKZSJGDQRLF-UHFFFAOYSA-N
prop-2-ynylcyclohexane
87265
97%

Chemical Identifiers

17715-00-3
122.211
UARFKZSJGDQRLF-UHFFFAOYSA-N
87265
C#CCC1CCCCC1
C9H14
MFCD00041562
3-cyclohexyl-1-propyne, prop-2-yn-1-ylcyclohexane, 1-prop-2-ynylcyclohexane, 3-cyclohexylpropyne, 2-propynylcyclohexane, propyne, 3-cyclohexyl, cyclohexane, 2-propynyl-9ci, prop-2-ynyl-cyclohexane, cyclohexane, 2-propynyl, 3-cyclohexylprop-1-yne
prop-2-ynylcyclohexane
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

RTECSNumber : UK4725000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only