Learn More
3-Cyclohexenecarboxylic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 294390010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Cyclohexenecarboxylic acid | |
4771-80-6 | |
100.0 | |
Blue-Gray to Brown-Yellow | |
238.0°C | |
Authentic | |
Glass bottle | |
1.4800 to 1.4820 | |
1 g | |
3-cyclohexene-1-carboxylic acid, 3-cyclohexenecarboxylic acid, cyclohex-3-enecarboxylic acid, 1,2,3,6-tetrahydrobenzoic acid, 1-cyclohexene-4-carboxylic acid, delta3-cyclohexenylcarboxylic acid, kyselina 1,2,5,6-tetrahydrobenzoova, 3-cyclohexene carboxylic acid, kyselina 1,2,5,6-tetrahydrobenzoova czech, .delta.3-cyclohexenecarboxylic acid | |
VUSWCWPCANWBFG-UHFFFAOYSA-N | |
cyclohex-3-ene-1-carboxylic acid | |
20903 | |
97% |
97% | |
96.0 | |
17.0°C | |
1.0800g/mL | |
>112°C | |
96% min. (GC) | |
C7H10O2 | |
MFCD00013781 | |
1.08 | |
Solubility in water: insoluble. Other solubilities: soluble in methanol | |
C1CC(CC=C1)C(=O)O | |
126.15 | |
126.15 | |
Liquid |
Chemical Identifiers
4771-80-6 | |
126.15 | |
VUSWCWPCANWBFG-UHFFFAOYSA-N | |
20903 | |
C1CC(CC=C1)C(=O)O |
C7H10O2 | |
MFCD00013781 | |
3-cyclohexene-1-carboxylic acid, 3-cyclohexenecarboxylic acid, cyclohex-3-enecarboxylic acid, 1,2,3,6-tetrahydrobenzoic acid, 1-cyclohexene-4-carboxylic acid, delta3-cyclohexenylcarboxylic acid, kyselina 1,2,5,6-tetrahydrobenzoova, 3-cyclohexene carboxylic acid, kyselina 1,2,5,6-tetrahydrobenzoova czech, .delta.3-cyclohexenecarboxylic acid | |
cyclohex-3-ene-1-carboxylic acid |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Danger
EINECSNumber : 225-314-7
RTECSNumber : GW3675000
TSCA : TSCA
RUO – Research Use Only