Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Cyclohexene-1-carboxaldehyde, 98%

Chemical Identifiers

Chemical Name or Material 3-Cyclohexene-1-carboxaldehyde
CAS 100-50-5
Molecular Formula C7H10O
Formula Weight 110.16g/mol
MDL Number MFCD00001572
UN Number UN2498
Beilstein 774001
Synonym 1,2,3,6-Tetrahydrobenzaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL04894AD

 
alfa aesar™
L04894-AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 163° to 164°C
Melting Point 1° to 3°C
Quantity 50mL
Flash Point 57°C (134°F)
Chemical Name or Material 3-Cyclohexene-1-carboxaldehyde
CAS 100-50-5
Assay 98%
Molecular Formula C7H10O
Formula Weight 110.16g/mol
MDL Number MFCD00001572
UN Number UN2498
Beilstein 774001
Synonym 1,2,3,6-Tetrahydrobenzaldehyde
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H226-H332-H319-H335
Toxic in contact with skin.
Flammable liquid and vapor.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: 1,2,3,6-TETRAHYDROBENZALDEHYDE
RTECS Number GW2800000
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H311-H226-H332-H319-H335
Toxic in contact with skin.
Flammable liquid and vapor.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.