Please login to your online account to display your discounted pricing

3-Cyclohexene-1,1-dimethanol, 98%, ACROS Organics™


Quantity 100g

Manufacturer: thermo fisher scientific  AC171831000

 View more versions of this product

Catalog No. AC171831000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Fine Crystalline Powder
Formula Weight 142.2
Quantity 100g
Color Beige
Beilstein 06, III, 4131
Melting Point 88° to 92°C
CAS Max % 100.0
Infrared Spectrum Authentic
Name Note 98%
Assay Percent Range 98%
Molecular Formula C8H14O2
Linear Formula C6H8(CH2OH)2
Chemical Name or Material 3-Cyclohexene-1, 1-dimethanol
MDL Number MFCD00001575
CAS 2160-94-3
Synonym 4, 4-Bis(hydroxymethyl)-1-cyclohexene
CAS Min % 97.5

Specifications

Chemical Name or Material 3-Cyclohexene-1, 1-dimethanol
Name Note 98%
CAS 2160-94-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H14O2
Linear Formula C6H8(CH2OH)2
Formula Weight 142.2
MDL Number MFCD00001575
Synonym 4, 4-Bis(hydroxymethyl)-1-cyclohexene