missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Cyclohexene-1,1-dimethanol, 98%, ACROS Organics™

Chemical Identifiers

CAS 2160-94-3
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C8H14O2
Linear Formula C6H8(CH2OH)2
Formula Weight 142.2
MDL Number MFCD00001575
Synonym 4, 4-Bis(hydroxymethyl)-1-cyclohexene
Chemical Name or Material 3-Cyclohexene-1, 1-dimethanol, 98%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC171831000 SDS Thermo Fisher Scientific
AC171831000
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

2160-94-3
100.0
C8H14O2
142.2
4, 4-Bis(hydroxymethyl)-1-cyclohexene
≥97.5%
97.5
98%
C6H8(CH2OH)2
MFCD00001575
3-Cyclohexene-1, 1-dimethanol, 98%

Specifications

2160-94-3
100.0
Fine Crystalline Powder
C8H14O2
142.2
06, III, 4131
Beige
100g
≥97.5%
97.5
Authentic
98%
C6H8(CH2OH)2
MFCD00001575
4, 4-Bis(hydroxymethyl)-1-cyclohexene
88° to 92°C
3-Cyclohexene-1, 1-dimethanol, 98%