missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Cyanoumbelliferone 98.0+%, TCI America™

$282.34 - $985.78

Chemical Identifiers

CAS 19088-73-4
Molecular Formula C10H5NO3
Molecular Weight (g/mol) 187.154
MDL Number MFCD00037480
InChI Key IJQYTHQDUDCJEQ-UHFFFAOYSA-N
Synonym 3-Cyano-7-hydroxycoumarin
PubChem CID 5393173
IUPAC Name 7-hydroxy-2-oxochromene-3-carbonitrile
SMILES C1=CC2=C(C=C1O)OC(=O)C(=C2)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C28341G View Documents Tci America
C28341G
1g Each for $282.34
 
C28345G View Documents Tci America
C28345G
5g Each for $985.78
 
Specifications

Chemical Identifiers

19088-73-4
187.154
IJQYTHQDUDCJEQ-UHFFFAOYSA-N
5393173
C1=CC2=C(C=C1O)OC(=O)C(=C2)C#N
C10H5NO3
MFCD00037480
3-Cyano-7-hydroxycoumarin
7-hydroxy-2-oxochromene-3-carbonitrile

Specifications

Yellow
1g
C10H5NO3
3-Cyano-7-hydroxycoumarin
C1=CC2=C(C=C1O)OC(=O)C(=C2)C#N
187.154
187.15
≥98.0% (GC)
280°C
19088-73-4
MFCD00037480
IJQYTHQDUDCJEQ-UHFFFAOYSA-N
7-hydroxy-2-oxochromene-3-carbonitrile
5393173
Crystal-Powder at 20°C
3-Cyanoumbelliferone
Safety and Handling

Safety and Handling

TSCA : No