missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Cyanopyridine-5-boronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H54385MD

 View more versions of this product

Catalog No. AAH54385MD



Specifications

Specifications

3-Cyanopyridine-5-boronic acid pinacol ester
250mg
C12H15BN2O2
3-cyano-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CN=C2)C#N
230.074
230.07
122°C to 124°C
402718-29-0
MFCD07780755
BOIKCRIMIQAFQJ-UHFFFAOYSA-N
5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carbonitrile
16414216
96%

Chemical Identifiers

402718-29-0
230.074
BOIKCRIMIQAFQJ-UHFFFAOYSA-N
16414216
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CN=C2)C#N
C12H15BN2O2
MFCD07780755
3-cyano-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine
5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carbonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents