Please login to your online account to display your discounted pricing

3-Cyanopropyltrichlorosilane, 97%, ACROS Organics™


Quantity 100mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC277111000

 View more versions of this product

Catalog No. AC277111000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 202.54
Physical Form Liquid
Assay ≥96.0%
Quantity 100mL
Color Clear colorless
Density 1.3000g/mL
CAS Max % 100.0
CAS Min % 96.0
Linear Formula CN(CH2)3Si(Cl)3
Synonym 4-(Trichlorosilyl)butyronitrile
CAS 1071-27-8
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Reacts violently with water.
Contact with water liberates toxic gas.
Chemical Name or Material 3-Cyanopropyltrichlorosilane
Molecular Formula C4H6Cl3NSi
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4620 to 1.4640 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material 3-Cyanopropyltrichlorosilane
CAS 1071-27-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4H6Cl3NSi
Linear Formula CN(CH2)3Si(Cl)3
Formula Weight 202.54
Synonym 4-(Trichlorosilyl)butyronitrile
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Reacts violently with water.
Contact with water liberates toxic gas.

SDS & Safety Documents