Please login to your online account to display your discounted pricing

3-Cyanopropyltrichlorosilane, 97%, ACROS Organics™

$64.87 - $64.87

Chemical Identifiers

Chemical Name or Material 3-Cyanopropyltrichlorosilane
CAS 1071-27-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4H6Cl3NSi
Linear Formula CN(CH2)3Si(Cl)3
Formula Weight 202.54
Synonym 4-(Trichlorosilyl)butyronitrile
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC277111000

 
thermo fisher scientific
AC277111000
Glass bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC277110250

 
thermo fisher scientific
277110250
Glass bottle 25mL Each for $64.87
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥96.0%
Formula Weight 202.54
Quantity 100mL
Color Clear colorless
Density 1.3000g/mL
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 3-Cyanopropyltrichlorosilane
Linear Formula CN(CH2)3Si(Cl)3
CAS 1071-27-8
Synonym 4-(Trichlorosilyl)butyronitrile
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Reacts violently with water.
Contact with water liberates toxic gas.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.4620 to 1.4640 (20°C, 589nm)
Assay Percent Range 97%
Molecular Formula C4H6Cl3NSi
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Reacts violently with water.
Contact with water liberates toxic gas.