Please login to your online account to display your discounted pricing

3-Cyanopropyltrichlorosilane, 97%, ACROS Organics™


Quantity 25mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  277110250

 View more versions of this product

Catalog No. AC277110250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 202.54
Quantity 25mL
Color Clear colorless
Density 1.3000g/mL
CAS Min % 96.0
CAS Max % 100.0
CAS 1071-27-8
Chemical Name or Material 3-Cyanopropyltrichlorosilane
Refractive Index 1.4620 to 1.4640 (20°C, 589nm)
Assay Percent Range 97%
Molecular Formula C4H6Cl3NSi
Linear Formula CN(CH2)3Si(Cl)3
Synonym 4-(Trichlorosilyl)butyronitrile
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Reacts violently with water.
Contact with water liberates toxic gas.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material 3-Cyanopropyltrichlorosilane
CAS 1071-27-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4H6Cl3NSi
Linear Formula CN(CH2)3Si(Cl)3
Formula Weight 202.54
Synonym 4-(Trichlorosilyl)butyronitrile
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Reacts violently with water.
Contact with water liberates toxic gas.

SDS & Safety Documents