missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Cyanopropyldimethylchlorosilane, 95%, ACROS Organics™

Chemical Identifiers

CAS 18156-15-5
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C6H12ClNSi
Linear Formula (CH3)2Si(Cl)(CH2)3CN
Formula Weight 161.7
Chemical Name or Material 3-Cyanopropyldimethylchlorosilane
Assay ≥94%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC313380500 SDS Thermo Fisher Scientific
AC313380500
Glass bottle 50mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC313380100 SDS Thermo Fisher Scientific
AC313380100
Glass bottle 10mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

18156-15-5
100.0
C6H12ClNSi
161.7
≥94%
94.0
95%
(CH3)2Si(Cl)(CH2)3CN
3-Cyanopropyldimethylchlorosilane

Specifications

0.9900g/mL
50mL
94.0
Authentic
1.4450 to 1.4470 (20°C, 589nm)
95%
(CH3)2Si(Cl)(CH2)3CN
3-Cyanopropyldimethylchlorosilane
Clear colorless to straw to brown
18156-15-5
100.0
Glass bottle
Liquid
C6H12ClNSi
161.7
≥94%

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger